(S)-Methyl-2,3-dihydroxypropanoate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2,3-dihydroxypropanoate

A113430-250MG A113430-1G A113430-10G
103798-232EA 20 USD
103798-232 103824-074 77025-878 77310-888 77570-734
(S)-Methyl-2,3-dihydroxypropanoate 97%
(S)-Methyl-2,3-dihydroxypropanoate

Precaución: for Research Use Only

Formula: C₄H₈O₄
MW: 120.1039 g/mol
Boiling Pt: 241.5 °C (760 mm Hg)
Flash Pt: 103.748 °C
Storage Temperature: Freezer
MDL Number: MFCD14636488
CAS Number: 10303-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR