(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(3,4-dihydroxyphenyl)propanoate 97%

Supplier: Ambeed
Warning

A204902-1G A204902-25G A204902-5G A204902-10G A204902-250MG
103817-576EA 12 USD
103817-576 103848-242 77086-436 77086-438 77086-440 77164-726 77164-728
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(3,4-dihydroxyphenyl)propanoate 97%
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-(3,4-dihydroxyphenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₆
MW: 311.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20260285
CAS Number: 37169-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR