(S)-Methyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate, Fmoc-Ser-OMe

A473598-10G A473598-250MG A473598-1G A473598-25G A473598-5G A473598-100G
103828-052EA 6 USD
103828-052 103839-126 76829-928 76829-930 76843-044 76843-046 76843-048 77572-488
(S)-Methyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate 97%
(S)-Methyl-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₅
MW: 341.363 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672334
CAS Number: 82911-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR