Fmoc-Tyr-OMe 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Methyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-hydroxyphenyl)propanoate
A175473-100G
A175473-10G
A175473-250MG
A175473-25G
A175473-5G
A175473-1G
103807-042EA
7
USD
103807-042
103845-350
76985-620
76985-622
76985-624
77082-694
77082-696
77301-374
Fmoc-Tyr-OMe 95%
Fmoc-Tyr-OMe
Precaución: for Research Use Only
Formula:
C₂₅H₂₃NO₅ MW: 417.4 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11044348 CAS Number: 82911-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...