(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 98%
Supplier: Ambeed
|
Warning
|
A182774-25G
A182774-5G
A182774-100G
A182774-10G
A182774-500G
103846-122EA
8
USD
103846-122
77083-680
77083-682
77138-776
77138-778
77138-780
77138-782
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid 98%
(S)-(+)-2-(4-Isobutylphenyl)propionic Acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₈O₂ Boiling Pt: 319.6±11.0 °C (760 mm Hg) Melting Pt: 49…52 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00069289 CAS Number: 51146-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...