(S)-(+)-Ethyl-3-piperidinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl L-nipecotate, Ethyl (S)-(+)-3-Piperidinecarboxylate, (S)-Ethyl piperidine-3-carboxylate

A117810-5G A117810-25G A117810-10G A117810-100G A117810-500G
103798-936EA 5 USD
103798-936 103824-828 77311-604 77566-150 77571-066 77610-968 77670-658
(S)-(+)-Ethyl-3-piperidinecarboxylate 98%
(S)-(+)-Ethyl 3-piperidinecarboxylate

Precaución: For research use only.

Formula: C₈H₁₅NO₂
Flash Pt: 76.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00792499
CAS Number: 37675-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR