(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

A185510-250MG A185510-5G A185510-1G
103815-858EA 6 USD
103815-858 103846-426 76872-874 77084-060 77301-980
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate 97%
(S)-Di-tert-butyl-5-oxopyrrolidine-1,2-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD03094770
CAS Number: 91229-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR