(-)-Dimethyl-L-malate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-(-)-Dimethyl Malate, (S)-Dimethyl 2-hydroxysuccinate, Dimethyl (S)-(-)-malate
A309729-25G
A309729-5G
A309729-10G
103835-778EA
5
USD
103835-778
76838-546
76838-548
77114-586
(-)-Dimethyl-L-malate 98%
(-)-Dimethyl-L-malate
Precaución: for Research Use Only
Formula:
C₆H₁₀O₅ Boiling Pt: 249.6 °C (760 mm Hg) Flash Pt: 55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066215 CAS Number: 617-55-0 UN: 2924 ADR: 3,8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...