Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 97%
Supplier: Ambeed
|
Warning
|
A128303-1000G
A128303-100G
A128303-10G
A128303-500G
A128303-25G
A128303-5G
103801-102EA
5
USD
103801-102
76811-038
76811-040
76811-042
76811-044
77313-408
77313-410
77313-412
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 97%
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate
Precaución: for Research Use Only
Formula:
C₁₂H₂₁NO₆ Boiling Pt: 370.9 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD03094774 CAS Number: 59279-60-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...