(S)-(+)-1,3-Butanediol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-1,3-Butylene glykol, (S)-Butane-1,3-diol, (S)-(+)-1,3-Dihydroxy butane

A233317-1G A233317-25G A233317-5G A233317-100G A233317-10G
103810-882EA 12 USD
103810-882 103850-642 77089-594 77089-596 77089-598 77140-114 77140-116
(S)-(+)-1,3-Butanediol 97%
(S)-(+)-1,3-Butanediol

Precaución: for Research Use Only

Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 207 °C (760 mm Hg)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00064278
CAS Number: 24621-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR