(S)-N-Boc-3-Amino-3-phenyl-1-propanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Boc-3-Amino-3-phenylpropan-1-ol

A102542-25G A102542-1G
103796-384EA 40 USD
103796-384 103822-080 77188-618 77308-960
(S)-N-Boc-3-Amino-3-phenyl-1-propanol 97%
(S)-N-Boc-3-Amino-3-phenyl-1-propanol

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₃
MW: 251.321 g/mol
Boiling Pt: 404.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09025325
CAS Number: 718611-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR