1-Benzyloxycarbonyl-(S)-3-methylpiperazine 96%

Supplier: Ambeed
Warning

Synonyms: Benzyl (3S)-3-methylpiperazine-1-carboxylate, (S)-Benzyl 3-methylpiperazine-1-carboxylate

A143287-100MG A143287-250MG A143287-1G A143287-5G
103803-262EA 33 USD
103803-262 77235-088 77235-090 77316-516 77316-518
1-Benzyloxycarbonyl-(S)-3-methylpiperazine 96%
1-Benzyloxycarbonyl-(S)-3-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.2942 g/mol
Boiling Pt: 358 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04039319
CAS Number: 612493-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR