(S)-Benzyl 3-aminopiperidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A100077-10G A100077-250MG A100077-1G A100077-5G
103795-236EA 16 USD
103795-236 103820-956 77188-512 77188-514 77282-424 77282-426
(S)-Benzyl 3-aminopiperidine-1-carboxylate 97%
(S)-Benzyl 3-aminopiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MW: 234.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08061980
CAS Number: 876461-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR