H-Glu(OBzl)-NCA 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Benzyl 3-(2,5-dioxooxazolidin-4-yl)propanoate

A333325-250MG A333325-1G A333325-10G A333325-100G
103825-632EA 14 USD
103825-632 103836-598 77032-924 77306-448 77568-676 77571-964
H-Glu(OBzl)-NCA 97%
H-Glu(OBzl)-NCA

Precaución: For research use only.

Formula: C₁₃H₁₃NO₅
Melting Pt: 97…98 °C
Storage Temperature: Freezer
MDL Number: MFCD11046651
CAS Number: 3190-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR