(S)-Benzyl 2-methylpiperazine-1-carboxylate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A101657-25G
A101657-5G
103796-070EA
40
USD
103796-070
103821-766
76982-184
77297-184
(S)-Benzyl 2-methylpiperazine-1-carboxylate hydrochloride 97%
(S)-Benzyl 2-methylpiperazine-1-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₉ClN₂O₂ MW: 270.76 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08436162 CAS Number: 1217720-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...