H-Lys(Boc)-OBzl·HCl 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Benzyl 2-amino-6-((tert-butoxycarbonyl)amino)hexanoate hydrochloride

A245844-1G A245844-100G A245844-10G A245844-250MG A245844-500G A245844-25G A245844-5G
103811-672EA 11 USD
103811-672 103851-500 77090-774 77153-184 77153-186 77153-188 77153-190 77304-366 77304-368
H-Lys(Boc)-OBzl·HCl 97%
H-Lys(Boc)-OBzl·HCl

Precaución: for Research Use Only

Formula: C₁₈H₂₈N₂O₄·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD11045868
CAS Number: 133170-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR