(S)-Benzyl-2,6-diaminohexanoate bis(4-methylbenzenesulfonate) 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Lys-OBzl·2p-TosOH, (S)-Benzyl 2,6-diaminohexanoate bis(4-methylbenzenesulphonate), H-Lys-OBzl·2 p-tosylate
A114260-10G
A114260-250MG
A114260-25G
A114260-1G
A114260-5G
103798-380EA
19
USD
103798-380
103824-220
77160-886
77160-888
77160-890
77297-300
77297-302
(S)-Benzyl-2,6-diaminohexanoate bis(4-methylbenzenesulfonate) 97%
(S)-Benzyl-2,6-diaminohexanoate bis(4-methylbenzenesulfonate)
Precaución: for Research Use Only
Formula:
C₂₇H₃₆N₂O₈S₂ MW: 580.711 g/mol Boiling Pt: 781.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08458627 CAS Number: 16259-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...