(S)-Benzyl 2-(2,5-dioxooxazolidin-4-yl)acetate 97%

Supplier: Ambeed
Warning

A663741-25G A663741-10G
103840-458EA 43 USD
103840-458 76844-652 77198-540
(S)-Benzyl 2-(2,5-dioxooxazolidin-4-yl)acetate 97%
(S)-Benzyl 2-(2,5-dioxooxazolidin-4-yl)acetate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₅
MW: 249.21 g/mol
Melting Pt: 120…122 °C
Storage Temperature: Freezer
MDL Number: MFCD16037555
CAS Number: 13590-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR