(S)-β-(Carbobenzoxyamino)-γ-butyrolactone 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-Benzyl (5-oxotetrahydrofuran-3-yl)carbamate, Benzyl (S)-Tetrahydro-5-oxo-3-furanylcarbamate, (S)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (S)-beta-(Cbz-amino)-gamma-butyrolactone, (S)-4-(Cbz-amino)tetrahydrofuran-2-one
A104813-100MG
A104813-10G
A104813-250MG
A104813-1G
A104813-5G
103796-782EA
10
USD
103796-782
103822-506
76687-196
76687-198
76687-200
77309-364
77309-366
(S)-β-(Carbobenzoxyamino)-γ-butyrolactone 97%
(S)-beta-(Carbobenzoxyamino)-gamma-butyrolactone
Precaución: for Research Use Only
Formula:
C₁₂H₁₃NO₄ MW: 235.24 g/mol Boiling Pt: 466 °C (760 mm Hg) Melting Pt: 103…105 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00216569 CAS Number: 87219-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...