(S)-Benzyl (4-(2,5-dioxooxazolidin-4-yl)butyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N6-Carbobenzoxy-L-lysine N-carboxyanhydride

A461533-25G A461533-250MG A461533-1G A461533-5G
103827-926EA 11 USD
103827-926 103839-004 76842-886 76990-824 77307-426 77307-428
(S)-Benzyl (4-(2,5-dioxooxazolidin-4-yl)butyl)carbamate 95%
(S)-Benzyl (4-(2,5-dioxooxazolidin-4-yl)butyl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₅
MW: 306.31 g/mol
Melting Pt: 120…124 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16036317
CAS Number: 1676-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR