(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-L-homoserine Lactone, (S)-alpha-(Carbobenzoxyamino)-gamma-butyrolactone, Cbz-Hse-Lactone, (S)-alpha-(Benzyloxycarbonylamino)-gamma-butyrolactone, N-Benzyloxycarbonyl-L-homoserine Lactone, (S)-Benzyl (2-oxotetrahydrofuran-3-yl)carbamate, N-Carbobenzoxy-L-homoserine Lactone, (S)-alpha-(Cbz-amino)-gamma-butyrolactone

A173115-10G A173115-1G A173115-25G A173115-5G
103806-792EA 24 USD
103806-792 103845-086 76720-582 77082-312 77082-314 77082-316
(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate 95%
(S)-Benzyl(2-oxotetrahydrofuran-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MW: 235.2400 g/mol
Melting Pt: 127…132 °C
Flash Pt: 235.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00674555
CAS Number: 35677-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR