N-Benzyloxycarbonyl-L-alaninol 97%

Supplier: Ambeed
Warning

Synonyms: Z-Ala-ol, (S)-Benzyl (1-hydroxypropan-2-yl)carbamate, (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester, N-Cbz-L-Alaninol

A110927-100G A110927-10G A110927-25G A110927-1G A110927-5G
103797-754EA 8 USD
103797-754 103823-562 76809-926 76809-928 76809-930 77310-364 77310-366
N-Benzyloxycarbonyl-L-alaninol 97%
N-Cbz-L-Alaninol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
Boiling Pt: 375.2 °C (760 mm Hg)
Melting Pt: 81…84 °C
Flash Pt: 180.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00672530
CAS Number: 66674-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR