L-Azetidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: L-Trimethyleneimine-2-carboxylic Acid, (S)-Azetidine-2-carboxylic acid, (S)-(-)-2-Azetidinecarboxylic acid, L-Azetidine-2-carboxylic Acid [Antagonist of L-Proline]

A391590-250MG A391590-1G A391590-25G A391590-5G A391590-10G
103799-642EA 18 USD
103799-642 103826-886 103837-924 76829-146 76841-316 76841-318 76841-320 77613-180
L-Azetidine-2-carboxylic acid 98%
L-Azetidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₇NO₂
MW: 101.11 g/mol
Melting Pt: 217 °C
Storage Temperature: Ambient
MDL Number: MFCD00005166
CAS Number: 2133-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR