Levofloxacin Q-acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Levofloxacin carboxylic acid, (S)-9,10-Difluoro-3-methyl-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid, (S)-(-)-9,10-Difluoro-2,3-dihydro-3-methyl-7-oxo-7H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylic Acid
A111727-10G
A111727-250MG
A111727-1G
A111727-5G
A111727-25G
A111727-100G
103797-894EA
22
USD
103797-894
103823-712
76718-106
76718-108
77284-172
77310-524
77599-606
77666-628
Levofloxacin Q-acid 97%
Levofloxacin Q-acid
Precaución: for Research Use Only
Formula:
C₁₃H₉F₂NO₄ MW: 281.21 g/mol Boiling Pt: 459.2 °C (760 mm Hg) Melting Pt: >300 °C Flash Pt: 231.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039905 CAS Number: 100986-89-8 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...