(S)-7-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: L-7-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

A208825-10G A208825-250MG A208825-1G A208825-5G
103817-888EA 32 USD
103817-888 103848-576 76691-394 76691-396 77303-072 77303-074
(S)-7-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid 97%
(S)-7-Hydroxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.202 g/mol
Boiling Pt: 456.3 °C (760 mm Hg)
Melting Pt: 273…275 °C
Flash Pt: 229.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00144534
CAS Number: 128502-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR