(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide 97%
Supplier: Ambeed
|
Warning
|
A106265-1G
A106265-5G
103797-020EA
11
USD
103797-020
103822-772
77283-462
77283-464
(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide 97%
(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide
Precaución: for Research Use Only
Formula:
C₆H₆ClNO₃S₂ Boiling Pt: 427.885 °C (760 mm Hg) Flash Pt: 212.578 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09033311 CAS Number: 160982-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...