H-Glu(OBzl)-OtBu·HCl 97%

Supplier: Ambeed
Warning

Synonyms: (S)-5-Benzyl 1-tert-butyl 2-aminopentanedioate hydrochloride, H-Glu(OBzl)-OtBu·HCl

A107301-100G A107301-10G A107301-1G A107301-5G A107301-25G
103797-164EA 8 USD
103797-164 103822-932 77148-528 77148-530 77283-570 77283-572 77549-064
H-Glu(OBzl)-OtBu·HCl 97%
H-Glu(OBzl)-OtBu·HCl

Precaución: for Research Use Only

Formula: C₁₆H₂₄ClNO₄
MW: 329.821 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038897
CAS Number: 105590-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR