Phenylacetylglutamine 96%

Supplier: Ambeed
Warning

Synonyms: (S)-5-Amino-5-oxo-2-(2-phenylacetamido)pentanoic acid, Phenylac-Gln-OH

A199725-250MG A199725-1G A199725-5G
103817-128EA 27 USD
103817-128 103847-754 76825-528 77085-802 77085-804
Phenylacetylglutamine 96%
Phenylacetylglutamine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₄
MW: 264.281 g/mol
Boiling Pt: 655.5 °C (760 mm Hg)
Melting Pt: 85…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01725188
CAS Number: 28047-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR