Hesperetin 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3',5,7-Trihydroxy-4'-methoxyflavanone, 4'-Methoxy-3',5,7-trihydroxyflavanone, (S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (±)-Hesperetin
A238234-100G
A238234-10G
A238234-500G
A238234-50MG
A238234-5G
A238234-1G
A238234-25G
103811-212EA
6
USD
103811-212
77098-402
77098-404
77098-406
77098-408
77098-410
77304-100
77304-102
Hesperetin 97%
Hesperetin
Precaución: for Research Use Only
Formula:
C₁₆H₁₄O₆ Boiling Pt: 586.2 °C (760 mm Hg) Melting Pt: 227…232 °C Flash Pt: 223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075646 CAS Number: 520-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...