(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected 98%

Supplier: Ambeed
Warning

Synonyms: (6S)-5-(tert-Butoxycarbonyl)-5-azaspiro[2.4]heptane-6-carboxylic acid, (S)-5-(tert-Butoxycarbonyl)-5-azaspiro[2.4]heptane-6-carboxylic acid

A275446-100G A275446-10G A275446-1G A275446-25G A275446-5G
103813-252EA 14 USD
103813-252 103853-178 76816-308 76816-310 77093-030 77093-032 77305-192
(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected 98%
(6S)-5-Azaspiro[2.4]heptane-6-carboxylic acid, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄
Boiling Pt: 377.4±35.0 °C (760 mm Hg)
Flash Pt: 182.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11099673
CAS Number: 1129634-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR