(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone 95%

Supplier: Ambeed
Warning

Synonyms: (5S)-(+)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-5-(Hydroxymethyl)dihydrofuran-2(3H)-one, (S)-(+)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone, (S)-(+)-5-Hydroxymethyltetrahydro-2-furanone

A437432-1G A437432-5G A437432-250MG A437432-25G
103827-622EA 37 USD
103827-622 103838-688 76842-426 76842-428 77168-740 77684-208
(S)-(+)-γ-Hydroxymethyl-γ-butyrolactone 95%
(S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone

Precaución: For research use only.

Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 308.9 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00066224
CAS Number: 32780-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR