(S)-5-(Aminomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one 98%

Supplier: Ambeed
Warning

A327000-100G A327000-100MG A327000-250MG A327000-1G A327000-25G A327000-5G
103825-452EA 8 USD
103825-452 103836-404 76817-118 76817-120 76817-122 76839-376 76839-378 76839-380
(S)-5-(Aminomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one 98%
(S)-5-(Aminomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₄H₁₈FN₃O₃
MW: 295.3 g/mol
Boiling Pt: 480 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD18379308
CAS Number: 168828-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR