(4S)-4-Methyloxazolidine-2,5-dione 95%

Supplier: Ambeed
Warning

A399797-1G A399797-5G A399797-10G A399797-250MG
103827-038EA 15 USD
103827-038 103838-072 76841-516 76841-518 77195-922 77195-924
(4S)-4-Methyloxazolidine-2,5-dione 95%
(4S)-4-Methyloxazolidine-2,5-dione

Precaución: for Research Use Only

Formula: C₄H₅NO₃
Storage Temperature: Freezer
MDL Number: MFCD03411276
CAS Number: 2224-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR