N-Methyl-L-leucine 98%

Supplier: Ambeed
Warning

Synonyms: (S)-4-Methyl-2-(methylamino)pentanoic acid, N-Me-Leu-OH

A223503-10G A223503-25G
103819-042EA 21 USD
103819-042 103849-922 76999-994 76999-996 77303-652
N-Methyl-L-leucine 98%
N-Methyl-L-leucine

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂
MW: 145.202 g/mol
Boiling Pt: 226.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00065919
CAS Number: 3060-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR