(S)-(-)-4-Isopropyl-2-oxazolidinone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one
A178286-25G
A178286-5G
A178286-100G
A178286-10G
A178286-500G
A178286-1G
103807-312EA
6
USD
103807-312
103845-652
77083-060
77083-062
77287-064
77287-066
77287-068
77301-552
(S)-(-)-4-Isopropyl-2-oxazolidinone 98%
(S)-(-)-4-Isopropyl-2-oxazolidinone
Precaución: for Research Use Only
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Boiling Pt: 291.1 °C (760 mm Hg) Melting Pt: 70…73 °C Flash Pt: 129.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010847 CAS Number: 17016-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...