(1S)-1-Amino-4-fluoroindane hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(1S)-2,3-Dihydro-4-fluoro-1H-inden-1-amine hydrochloride, (S)-4-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride
A130665-100MG
A130665-250MG
A130665-1G
103801-450EA
276
USD
103801-450
76972-048
76972-050
77298-420
(1S)-1-Amino-4-fluoroindane hydrochloride 95%
(1S)-1-Amino-4-fluoroindane hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₀FN·ClH Storage Temperature: Ambient |
MDL Number:
MFCD16295140 CAS Number: 1286734-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...