(S)-4-Boc-morpholine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2S)-Morpholine-2-carboxylic acid, N-BOC protected, (S)-4-Boc-Morpholine-2-carboxylic acid

A108366-250MG A108366-25G A108366-5G
103797-338EA 22 USD
103797-338 103823-118 77188-936 77188-938 77283-714
(S)-4-Boc-morpholine-2-carboxylic acid 95%
(S)-4-Boc-morpholine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
MW: 231.2457 g/mol
Boiling Pt: 369.5 °C (760 mm Hg)
Melting Pt: 127…134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06200662
CAS Number: 868689-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR