(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone 97%

Supplier: Ambeed
Warning

Synonyms: (4S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone, (S)-4-Benzyl-3-propionyloxazolidin-2-one

A143862-100G A143862-10G A143862-25G A143862-1G A143862-5G
103803-330EA 6 USD
103803-330 76996-990 76996-992 77299-360 77316-636 77316-638
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone 97%
(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₃
Melting Pt: 44…46 °C
Flash Pt: 193.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00269688
CAS Number: 101711-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR