(S)-(-)-4-Amino-2-hydroxybutyric acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-4-Amino-2-hydroxybutanoic acid
A441449-25G
A441449-100G
A441449-10G
A441449-500G
A441449-5G
103838-742EA
5
USD
103838-742
76842-506
77279-120
77279-122
77279-124
77307-316
(S)-(-)-4-Amino-2-hydroxybutyric acid 97%
(S)-(-)-4-Amino-2-hydroxybutyric Acid
Precaución: for Research Use Only
Formula:
C₄H₉NO₃ Boiling Pt: 361.8 °C (760 mm Hg) Melting Pt: 207…212 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00674110 CAS Number: 40371-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...