4,4-Difluoro-L-prolinamide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-4,4-Difluoropyrrolidine-2-carboxamide hydrochloride

A183070-100MG A183070-250MG
103815-600EA 23 USD
103815-600 103846-164 77029-508 77029-510
4,4-Difluoro-L-prolinamide hydrochloride 97%
4,4-Difluoro-L-prolinamide hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈F₂N₂O·HCl
MW: 186.59 g/mol
Melting Pt: 221…223 °C
Storage Temperature: Ambient
MDL Number: MFCD06797014
CAS Number: 426844-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR