(S)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A589091-1G
A589091-5G
A589091-10G
A589091-250MG
A589091-25G
103828-914EA
13
USD
103828-914
103840-058
76844-200
76844-202
77197-872
77197-874
77307-902
(S)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid 97%
(S)-4-(tert-Butoxycarbonyl)morpholine-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₇NO₅ Boiling Pt: 369.5 °C (760 mm Hg) Melting Pt: 169…173 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114904 CAS Number: 783350-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...