(S)-4-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)butanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-β-Hse(Bzl)-OH

A105650-100MG A105650-250MG A105650-1G
103796-918EA 83 USD
103796-918 103822-656 77160-462 77160-464 77682-970
(S)-4-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)butanoic acid 95%
(S)-4-(Benzyloxy)-3-((tert-butoxycarbonyl)amino)butanoic acid

Precaución: For research use only.

Formula: C₁₆H₂₃NO₅
MW: 309.35 g/mol
Boiling Pt: 480.5±45.0 °C (760 mm Hg)
Flash Pt: 244.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08282798
CAS Number: 718608-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR