(S)-4-(4-Aminobenzyl)oxazolidin-2-one 97%

Supplier: Ambeed
Warning

Synonyms: (4S)-4-[(4-Aminophenyl)methyl]-2-oxazolidinone, (S)-4-(4-Aminobenzyl)-2(1H)-oxazolidinone

A387352-100G A387352-10G A387352-25G A387352-5G A387352-1G
103826-822EA 10 USD
103826-822 103837-864 76669-782 76669-784 76841-226 76841-228 77306-980
(S)-4-(4-Aminobenzyl)oxazolidin-2-one 97%
(S)-4-(4-Aminobenzyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Melting Pt: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD03411476
CAS Number: 152305-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR