(S)-4-Cyano-4'-(2-methylbutyl)biphenyl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-4'-(2-Methylbutyl)-[1,1'-biphenyl]-4-carbonitrile

A166721-250MG A166721-5G
103806-092EA 48 USD
103806-092 103844-354 77191-294 77300-846
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl 98%
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl

Precaución: Sealed in dry, at room temperature.

Formula: C₁₈H₁₉N
MW: 249.36 g/mol
Boiling Pt: 388.8 °C (760 mm Hg)
Flash Pt: 190.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00192280
CAS Number: 63799-11-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR