(S)-3-Methyl-pentanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Methylvaleric Acid

A539598-1G A539598-5G
103828-620EA 53 USD
103828-620 103839-742 76843-788 76843-790
(S)-3-Methyl-pentanoic acid 97%
(S)-3-Methyl-pentanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₂
MW: 116.158 g/mol
Boiling Pt: 199 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02262177
CAS Number: 1730-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR