(S)-3-Methyl-pyrrolidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Methylpyrrolidine hydrochloride

A131693-100MG A131693-250MG A131693-1G A131693-5G
103801-618EA 166 USD
103801-618 77109-282 77109-284 77314-128 77314-130
(S)-3-Methyl-pyrrolidine hydrochloride 97%
(S)-3-Methyl-pyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁N·ClH
Boiling Pt: 146.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06796632
CAS Number: 186597-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR