(S)-3-Methylpiperidine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A428995-1G
A428995-5G
A428995-100MG
A428995-10G
A428995-250MG
A428995-25G
103827-500EA
20
USD
103827-500
103838-556
76842-234
76842-236
77016-544
77016-546
77016-548
77016-550
(S)-3-Methylpiperidine hydrochloride 98%
(S)-3-Methylpiperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₄ClN Storage Temperature: Ambient |
MDL Number:
MFCD25541503 CAS Number: 155797-02-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...