(S)-3-Methoxypyrrolidine hydrochloride 95%

Supplier: Ambeed
Warning

A101259-100MG A101259-10G A101259-250MG A101259-1G A101259-5G
103795-874EA 23 USD
103795-874 103821-558 76780-482 76780-484 76780-486 77282-692 77282-694
(S)-3-Methoxypyrrolidine hydrochloride 95%
(S)-3-Methoxypyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD11849817
CAS Number: 685828-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR