(S)-3-Hydroxypiperidine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-3-Piperidinol hydrochloride
A128655-100G
A128655-250MG
A128655-1G
A128655-25G
A128655-5G
103801-170EA
6
USD
103801-170
76983-572
76983-574
77313-508
77313-510
77313-512
(S)-3-Hydroxypiperidine hydrochloride 98%
(S)-3-Hydroxypiperidine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO MW: 137.6100 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426348 CAS Number: 475058-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...