(S)-3-Hydroxybutyric acid 96%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Hydroxybutanoic acid, (S)-beta-Hydroxybutyric acid

A188212-100MG A188212-250MG A188212-1G A188212-5G
103799-456EA 31 USD
103799-456 103816-080 103846-670 77191-870 77191-872 77302-112 77302-114
(S)-3-Hydroxybutyric acid 96%
(S)-3-Hydroxybutyric acid

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₄H₈O₃
MW: 104.1 g/mol
Melting Pt: 45…50 °C
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00137685
CAS Number: 6168-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR